Learn More
Biomatik Corporation Goat Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit
Supplier:  Biomatik Corporation EKU07721
Encompass_Preferred
Goat Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit for detecting TIMP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A140C4Q7. Gene ID: 102190027. Target Synonyms: SFD; TIMP Metallopeptidase Inhibitor 3; Metallopeptidase Inhibitor 3; Sorsby Fundus Dystrophy; Pseudoinflammatory. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5571
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.