Learn More
ORS Nasco GOGGLE UVEX STRAT S3805 10/PK
Supplier:  ORS Nasco 763S3805
Encompass_Preferred
Honeywell part# S3805. Honeywell Uvex Strategy Safety Goggles Frame Color: Gray Includes: Neoprene band Ventilation Details: Closed vent Lens Tint: Clear Lens Type: Anti-fog Lens Coating: Uvextra Safety goggles Recommended Applications: Safety
Catalog No. 50-255-1573
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.