Learn More
3M™ GoggleGear™ 6000 Series Scotchgard™ Goggles
Catalog No. 19071100 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Black
Blue
Gray
Green
Red
Lens Tint:
Clear
Gray
Headband Material:
Neoprene
Nylon
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Headband Material
19-071-100 Gray Clear Neoprene
19-071-099 Black Gray Nylon
19-071-096 Black Clear Nylon
19-071-097 Green Clear Nylon
19-071-098 Red Clear Nylon
19-071-101 Blue Clear Neoprene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-071-100 Supplier 3M™ Supplier No. 7100191929
This item is not returnable. View return policy
3M™ Scotchgard™ Anti-Fog Coating helps to increase eyewear performance in wet and steamy environments.
  • Indirect venting allows air to circulate and helps reduce fogging
  • D3 (droplet and splash) and D4 (dust) rated
  • Polycarbonate lens absorbs 99.9% UVA and UVB and meet the requirements of ANSI Z87.1- 2015 Standard as an Impact Rated spectacle
  • 3M Catalog# GG6001AF-GRY
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Primary Metals, Chemical Handling, Painting, General Manufacturing, Oil and Gas
Frame Color Gray
Headband Material Neoprene
No. per Case 10/Cs.
Model GG6001AF-GRY
Size Universal
Ventilation Details Indirect venting
Material Polycarbonate
Lens Material Polycarbonate
UV Protection UV400