Learn More
Moldex™ Goin' Green™ PlugStation™ Ear Plug Dispenser
SDP
Goin' Green™ PlugStation™ dispensers are pre-filled with ear plugs and are convenient for any work environment. Easy access, wall-mountable unit will help ensure compliance. When the PlugStation is empty, simply dispose of it and hang a new one.
$636.50 - $843.50
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Easy Access to Hearing Protection for Workers and Facility Visitors in Reception Areas, Break Rooms, Workstations, and Other Noisy Environments.
Plug Material Foam
View More Specs
Products 2
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
19-160-0159
SDP
Accessories View Documents
Moldex™
6646
250 Pairs
Case of 1500 for $636.50
Only null left
Add to Cart
19-160-0160
SDP
Accessories View Documents
Moldex™
6647
500 Pairs
Case of 200 for $843.50
Only null left
Add to Cart
Description
Description
  • Holds Goin' Green™ disposable ear plugs
  • PlugStations are sealed to ensure ear plugs stay clean and sanitary
  • High visibility to increase compliance
  • Dispenses one pair at a time to eliminate wasteful grabbing
  • Includes mounting bracket
  • Available in two sizes - Small 250 Pair/Dispenser or Large 500 Pair/Dispenser
Specifications
Specifications
Yes
Universal
Foam
33 dB
Bullet
PVC-free Plastic
Mounting Hardware
Class AL
Easy Access to Hearing Protection for Workers and Facility Visitors in Reception Areas, Break Rooms, Workstations, and Other Noisy Environments.
Green
Uncorded
Single-use
Wall Mountable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.