Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Skin Lotion
SDP

Catalog No. 19053331 Shop All Gojo Industries, Inc. Products
Click to view available options
Volume (Metric):
147 mL

Non-greasy, silicone free moisturizing lotion that helps condition skin

  • Non-greasy, silicone free moisturizing lotion that helps condition skin
  • Contains vitamin E and aloe
TRUSTED_SUSTAINABILITY

Specifications

Product Type Skin Lotion
Color White
Form Lotion
For Use With (Application) Prevents chapping and rednes
Packaging Tube
Quantity 1 Ea.
Volume (English) 5 oz.
Volume (Metric) 147 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.