Learn More
Description
- Ideal for customer service, checkout and other locations
- Puts Purell™ LTX™ and TFX™ dispensers on any table or counter
- Dispenser sold separately
Specifications
Specifications
| Product Type | Tabletop Stand |
| Color | White |
| For Use With (Equipment) | Touch-free dispensers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
