Learn More
Medchemexpress LLC Gold(1+), dichloro(4,4′-dimethoxy-2,2′-bipyridine-κN1,κN1′)-, (SP-4-2)-, hexafluorophosphate(1-) | 1221591-26-9 | 99.0% | 629.07 g/mol | C12H12AuCl2F6N2O2P | 5 MG

Supplier: Medchemexpress LLC HY1388695MG
AubipyOMe is a gold-containing bipyridine complex used as a biochemical tool to inhibit tartrate-resistant acid phosphatase (TRAP/ACP5). It is supplied as a solid with high purity for research applications, enabling studies of osteoclast and macrophage TRAP activity and related biochemical assays.
- Gold-containing bipyridine complex.
- Potent TRAP/ACP5 inhibitor; IC50 = 1.3 μM (TRAP5a), 1.8 μM (TRAP5b).
- Molecular weight 629.07 g/mol; formula C12H12AuCl2F6N2O2P.
- Purity 99.0% by supplier analysis.
- Solid, off-white to light yellow appearance.
- Supplied in small research quantities, suitable for biochemical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.