Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Gold Chloride Trihydrate ACS, MP Biomedicals™

Catalog No. 0215252601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-152-52601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-152-52601 Supplier MP Biomedicals™ Supplier No. 0215252601
Add to Cart
Add to Cart

MP Biomedicals™ Gold Chloride Trihydrate ACS for microanalysis of rubidium, cesium and in the determination of alkaloids.

  • Used for microanalysis of rubidium, cesium and in the determination of alkaloids.
  • Usually used as a harbinger for the preparation of gold (Au) nanoparticles.
  • It is used in the control rods of nuclear reactors and in the manufacturing of electric light filaments.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
Synonym Tetrachlorogold (III) Hydrate
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Quantity 1 g
Formula Weight 393.8
Grade ACS
Chemical Name or Material Gold Chloride, Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.