CAS RN 16961-25-4
Tetrachloroauric(III) acid trihydrate, ACS reagent
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Gold Chloride Trihydrate, Certified ACS, Fisher Chemical™
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 MDL Number: MFCD00149904 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K Synonym: Gold(III) chloride trihydrate, Tetrachloroauric acid trihydrate, Hydrogren tetrachloroaurate(III), Tetrachloroauric(III) acid, Chloroauric acid trihydrate IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Gold Chloride, Trihydrate, ACS, 49%, Spectrum™ Chemical
CAS: 16961-25-4 Molecular Formula: AuCl4H5 Molecular Weight (g/mol): 343.81 InChI Key: QJXOQVKCBDMTLU-UHFFFAOYSA-O IUPAC Name: hydrogen tetrachlorogolduide SMILES: [H+].Cl.Cl.Cl.Cl.[Au-]
Gold Chloride Trihydrate ACS, MP Biomedicals™
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K Synonym: Tetrachlorogold (III) Hydrate IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Hydrogen tetrachloroaurate(III) trihydrate, ACS, 99.99% (metals basis), Au 49.0% min
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 MDL Number: MFCD00149904 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K Synonym: Chloroauric acid IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Tetrachloroauric(III) Acid Trihydrate, 99.5%, For Analysis, EMSURE™, MilliporeSigma™
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K Synonym: Gold(III)chloride hydrochloric acid,Gold trichloride hydrochloride IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Gold Chloride Trihydrate ACS, 5 g, MP Biomedicals™
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 393.83 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K Synonym: Tetrachlorogold (III) Hydrate IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Gold Chloride, 1% (w/v), Ricca Chemical
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Gold Chloride, 1% (w/v), LabChem™
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Gold AA Standard, Certified, 1000 ppm (1mL = 1mg Au), LabChem™
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Gold(III) chloride trihydrate, Solstice
CAS: 16961-25-4 Molecular Formula: AuCl4H Molecular Weight (g/mol): 339.77 MDL Number: MFCD00149904 InChI Key: VDLSFRRYNGEBEJ-UHFFFAOYSA-K IUPAC Name: trichlorogold hydrochloride SMILES: Cl.Cl[Au](Cl)Cl
Sigma Aldrich Gold(III) chloride trihydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.