Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold(I) chloride, 99+%

Catalog No. p-3523459
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388330010 1 g Glass bottle
AC388330050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC388330010 Supplier Thermo Scientific Chemicals Supplier No. 388330010

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

This item is not returnable. View return policy

CAS: 10294-29-8 | AuCl | 232.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-29-8
Molecular Formula AuCl
Molecular Weight (g/mol) 232.42
MDL Number MFCD00046175
InChI Key FDWREHZXQUYJFJ-UHFFFAOYSA-M
Synonym gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
PubChem CID 27366
ChEBI CHEBI:30078
IUPAC Name chlorogold
SMILES [Cl-].[Au+]

Specifications

Melting Point 289.0°C
Color Yellow to Green
Physical Form Powder
Quantity 1 g
Assay Percent Range 99+%
Linear Formula AuCl
Merck Index 15, 4550
Solubility Information Solubility in water: decomposes
Formula Weight 232.42
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Gold(I) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 233-655-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.