Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solstice Gold(III) chloride trihydrate, Solstice

Catalog No. 6000628 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-28 25 g
60-006-27 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-006-28 Supplier Solstice Supplier No. G402225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥49.0% Au basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149904
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Physical Form Solid
Quantity 25 g
Assay Percent Range ≥49.0% (Au basis)
Linear Formula HAuCl4 · 3H2O
UN Number UN3260
Synonym Hydrogen Tetrachloroaurate(III), Tetrachloroauric(III) Acid
PubChem CID 329799921
Formula Weight 393.83g/mol
Percent Purity ≥49% (Au basis)
Grade ACS
Concentration or Composition (by Analyte or Components) ≥49.0% Au
Manufacturing Origin USA
Packaging Glass Bottle
Product Line Solstice
Residue after Ignition ≤0.2% (as sulfates) max
Decomposition Information Chlorine Gas, Hydrogen Chloride, Metallic Gold
Chemical Name or Material Gold(III) chloride trihydrate
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
RTECSNumber MD5428000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.