Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Gold Leaf Electroscope
SDPSCI_ED

Catalog No. S139481 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139481 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139481 Supplier United Scientific™ Supplier No. ELGL01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The gold leaf electroscope is a sensitive electroscope type that is used for detecting charges.

Designed to detect the presence of electric charge, this sturdy electroscope features a metal case with front and rear glass windows to prevent air currents from affecting the deflection of the leaves.

  • Leaves are gold and aluminum
  • Leaf support rod is held in place by an insulating plastic sleeve
  • A graduated scale is also provided for measuring the deflection from 0 to 90 degrees.
  • Includes activity guide

Specifications

Grade 5 to 12
Format Model
Product Type Electroscope
Includes Electroscope and Activity Guide
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.