Learn More
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Description
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown to Pink |
| Quantity | 100 mL |
| Particle Size | 250nm |
| Sensitivity | Light sensitive |
| Form | Liquid |
| Percent Purity | 99% |
| Chemical Name or Material | Gold nanoparticles, supplied in 0.1mM PBS, reactant-free |
Safety and Handling
| TSCA | Yes |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.