Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold wire, 14kt, yellow, 1.63mm (0.064in) dia, Au 58.3% min

Catalog No. AA40329BS
Change view
Click to view available options
Quantity:
10 cm
25 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40329BS 10 cm
AA40329BU 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40329BS Supplier Thermo Scientific Chemicals Supplier No. 040329BS
Add to Cart

Gold wire is used in jewelry and as a plated coating on a wide variety of electrical and mechanical components. It is also acts as a heat reflecting coating for glass as well as a decorative medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Chemical Name or Material Gold wire (14kt)
Name Note 1.63mm (0.064 in.) dia.
Quantity 10 cm
Weight ≈28g/m
Form Wire
MDL Number MFCD00003436
Solubility Information Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water.
Concentration or Composition (by Analyte or Components) % Concentration Cu>Zn>Ag, Au ≤58.3%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.