Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golden Syrian Hamster IgG Isotype Control, HRP, Novus Biologicals™

Golden Syrian Hamster Antibody
Supplier: Novus Biologicals NBP196965
Description
IgG specifically detects IgG in Golden Syrian Hamster samples. It is validated for Western Blot, ELISA.Specifications
| IgG | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate | |
| IGHG1 | |
| Multi-step | |
| RUO | |
| 3500 | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| HRP | |
| Immunoglobulin G, ImmunoglobulinG | |
| Golden Syrian Hamster | |
| 1 mg | |
| Isotype Control (Primary) | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti- Hamster IgG and anti-Hamster Serum. | |
| Golden Syrian Hamster | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction