Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius GoldLINE Electrodes

Catalog No. 14558081 Shop All Sartorius Products
Change view
Click to view available options
Type:
Pharma grade
Speartip
Junction:
3X Hole Junction
3X Junction
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Junction
14-558-081 Speartip 3X Hole Junction
14-558-079 Pharma grade 3X Junction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-558-081 Supplier Sartorius Supplier No. PYP302S

Includes

Electrode, Manual, 50mL refill bottle

Superior stable results due to patented three chamber Iodine/Iodide metal-ion free reference system that lower temperature sensitivities and provides a metal ion free reference system ideal for Pharma and R&D grade application.

Up your performance and accuracy with GoldLINE. The unique three-chamber reference system is more stable with a metal-free iodide/iodine reference system compared to our excellent Ag/AgCl reference system found in our PlatiumLINE series. GoldLINE not only offers unbeatable stability and faster response times but higher accuracy and precision. Accelerated performance is attributed to the advanced patented reference technologies and superior glass and reference materials & construction. The GoldLINE is a superior sensor that gives precise results allowing for independence from sample contamination with minimal temperature influences.
  • GoldLINE electrodes have lower temperature sensitivity, reducing the drastic influences temperature variants can have on pH measurement, thus producing more precise results.
  • Exchangeable bridge electrolyte enables one to match the internal electrolyte to their sample for better measurements.
  • Speed and response time is quicker due to stability of reference exchange, system and damping of temperature influences and the reduction of metal or precipitant contamination that can occur with other electrodes.

Patented junction and three-chamber systems set sensors apart from the rest
  • Reduce and eliminate metal ion contamination and chelation
  • Minimizes damage to sensors elongating the life of the electrodes, saving time and money as well as increased productivity to mitigate faulty measurements

Smart ID sensor versions
  • A reliable and accurate measurement through automatic electrode recognition between sensor and meter
  • Provide unique identification for each sensor for traceability
  • Can be used on multiple instruments without having to recalibrate
  • Stores the last calibration of the sensor
  • Completes the documentation trail between sensor and meter for the highest GLP practices

  • Electrode includes model, serial number with date, time and measurement values


PY-P28-2S
Pharma Grade – pH/ATC, TRIS Impervious, Glass Body, Liquid Filled 3M KCl, 3X Junction, Iodine/Iodide reference

PY-P29-2S
Micro – 5mm dia., pH/ATC, TRIS Impervious, Glass Body, Liquid Filled 3M KCl, 3X Junction, Iodine/Iodide-reference

PY-P30-2S
Speartip pH/ATC Glass Body, 3x, Hole Junction, Polymer Filled

Warranty and Services

1 year

TRUSTED_SUSTAINABILITY

Specifications

Type Speartip
Material Glass
Junction 3X Hole Junction
Reference Type Iodine/Iodide
Connector Types BNC (2.5 mm Plug)
Storage Requirements Ambient temperature
Filling Solution Type Polymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.