Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLPH2 Recombinant Rabbit Monoclonal Antibody (015), FITC
GREENER_CHOICE

Catalog No. PIMA540949
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540949 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540949 Supplier Invitrogen™ Supplier No. MA540949
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi transmembrane protein. It processes protein synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus. The expression of this encoded protein has been observed to be upregulated in response to viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH2
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 015
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 0.5% BSA and 0.09% sodium azide
Gene GOLM1
Gene Accession No. Q8NBJ4
Gene Alias 2310001L02Rik; AW125446; bA379P1.3; C9orf155; D030064E01Rik; epididymis luminal protein 46; FLJ22634; FLJ23608; Golgi membrane protein 1; Golgi membrane protein GP73; golgi phosphoprotein 2; golgi protein, 73-kD; GOLM1; Golph2; GOLPH2Ê; GP73; HEL46; PSEC0242; PSEC0257; RP11-379P1.3; UNQ686/PRO1326
Gene Symbols GOLM1
Host Species Rabbit
Immunogen Recombinant Human GOLM1/GP73 protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51280
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.