Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLPH3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119861 Supplier Invitrogen™ Supplier No. PA5119861
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Hela cell lysate, Daudi cell lysate, MCF-7 cell lysate, SK-Br-3 cell lysate, rat ovary tissue lysate, rat large intestine tissue lysate, mouse colon tissue lysate, rat large intestine tissue, A549, Hela. Subcellular Location: Golgi stack membrane, trans-Golgi network membrane.

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is localized at the Golgi stack and may have a regulatory role in Golgi trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH3L
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene GOLPH3L
Gene Accession No. Q66H74, Q8R088, Q9H4A5
Gene Alias 2010204I15Rik; AU015439; AW549227; C77637; golgi phosphoprotein 3 like; golgi phosphoprotein 3-like; Golph3l; GPP34R; GPP34-related protein
Gene Symbols GOLPH3L
Host Species Rabbit
Immunogen Recombinant protein within human GOLPH3L aa 50-250.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229593, 310669, 55204
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.