Learn More
Apexbio Technology LLC Golvatinib (E7050) 928037-13-2 50mg
SDP
Supplier:  Apexbio Technology LLC B143950
Encompass_Preferred
Golvatinib (E7050 CAS 928037-13-2) is a small-molecule inhibitor targeting the tyrosine kinases c-Met and VEGFR-2 Golvatinib inhibits phosphorylation of c-Met in MKN45 gastric cancer cells and VEGFR-2 in human umbilical vein endothelial cells (HUVECs) with IC50 values of 14 nM and 16 nM respectively Additionally it suppresses proliferation of various tumor cells including MKN45 EBC-1 Hs746T and SNU-5 and inhibits tumor growth in xenograft models via reduction of c-Met and VEGFR-2-mediated pathways Golvatinib is utilized in cancer research to explore targeted antitumor efficacy related to angiogenesis and oncogenic signaling
Catalog No. 50-000-62067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.