Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Goniometer
SDPSCI_ED

Catalog No. S16160ND Shop All Vernier Software Products
Change view
Click to view available options
Range:
0 to 340°
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S16160ND 0 to 340°
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16160ND Supplier Vernier™ Supplier No. GNMBTA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Includes

Goniometer: joint angle sensor with base plate attached to static arm, 12-inch flexible joint arm, 10-inch flexible joint arm, 8-inch flexible joint arm, Two 24-inch elastic, hook-and-loop straps, Two 14-inch elastic, hook-and-loop straps

Requires

Requires an interface and software, sold separately

The Goniometer measures the angle of a joint, such as the knee or elbow. Use it to analyze the range of motion of a limb during different types of physical activity.

Range: 0-340° (+/-170°) Resolution: 13-bit Typical Resolution: 0.06° 12-bit Typical Resolution: 0.12° 10-bit Typical Resolution: 0.5°

Warranty and Services

5-year limited warranty

Specifications

Grade High School and College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Accuracy ±170°
Range 0 to 340°
Resolution 0.06° at 13-bit, 0.12° at 12-bit, 0.5° at 10-bit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.