Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Goodie Bone, Large
SDP

Catalog No. 14727012
Encompass
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
14-727-012 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-727-012 Supplier Bio-Serv™ Supplier No. K3536
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Made of flexible, natural rubber
  • Use with treats for added interest
  • Cagewasher safe
  • 8 in. long x 2 in. wide
  • Red

Specifications

Product Type Canine Enrichment Device
Color Red
Material Rubber
Size Large
Length (English) 8 in.
Width (English) 2 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.