Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Autoclave Bags Printed with Biohazard Logo

Catalog No. 07202043 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
3 L
9 L
14 L
33 L
Width (Metric):
255 mm
300 mm
400 mm
600 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Width (Metric)
07-202-043 3 L 255 mm
07-202-044 9 L 300 mm
07-202-045 14 L 400 mm
07-202-047 33 L 600 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-202-043 Supplier Corning™ Supplier No. SA0310
Add to Cart
Add to Cart

Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.

  • Polypropylene (PP) resistant bags
  • Thermal resistance 134°C
  • Transparent
TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
No. per Case 500/Cs.
No. per Pack 1/Pk.
Height (Metric) 400 mm
Capacity (Metric) 3 L
Width (Metric) 255 mm
Thickness (Metric) 40 μm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.