Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ gp120 (ZM53M.PB12) (HIV-1/Clade C) (AY423984, 46 a.a. - 490 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells

Catalog No. 89946300 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-300 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-946-300 Supplier Abnova™ Supplier No. P5111
Add to Cart
Add to Cart

Used for SDS-PAGE

Specifications

Accession Number AY423984
Concentration 1 ug/uL
For Use With (Application) SDS-PAGE
Formulation Liquid
Name gp120 (ZM53M.PB12) (HIV-1/Clade C) Recombinant Protein
Preparation Method Mammalian cell (293) expression system
Quality Control Testing SDS-PAGE Result
Quantity 50 μg
Storage Requirements Store at 4°C. Do not freeze. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name GP120(ZM53M.PB12)(HIV-1/CLADE
Species Human
Recombinant Recombinant
Protein Tag His
Expression System Mammalian cell (293) expression system
Form Liquid
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.