Learn More
ACROBiosystems Recombinant Protein;Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;GP0,GP120,HIV,GP160,HIV-1,GP41,GP120-CN54,CN54,CN54-GP120
Supplier: ACROBiosystems GP0V182E625UG
Recombinant Protein;Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120, His Tag, Avi Tag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession # G4XFJ5-1 (E46G, T396A, A497T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObiosystems.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 56.7 kDa. The protein migrates as 90-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 5 μg/mL (100 μL/well) can bind Biotinylated HIV-1 [HIV-1/Clade B/C (CN54)] GP120, His Tag, Avi Tag (Cat. No. GP0-V182E6) with a linear range of 0.156-1.25 μg/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.