Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542593 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542593 Supplier Invitrogen™ Supplier No. PA542593
  • $624.00
    $485.65 / Each of 1

    Save $138.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Peptide sequence: SLQAALQPIV SSLNVSVDGN GEFIVRMALF QDQNYTNPYE GDAVELSVES Sequence homology: Cow: 79%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 79%; Rat: 92%.

GP2 is component of pancreatic secretory (zymogen) granule. The exact function of GP2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GP2
Gene Accession No. P55259
Gene Alias 2310037I18Rik; AV060639; glycoprotein 2; glycoprotein 2 (zymogen granule membrane); glycoprotein 80; GP2; gp80; Pancreatic secretory granule membrane major glycoprotein GP2; pancreatic zymogen granule membrane associated protein GP2; pancreatic zymogen granule membrane protein GP-2; secretory (zymogen) granule membrane glycoprotein GP2; ZAP75; zymogen granule membrane glycoprotein 2
Gene Symbols GP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GP2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2813
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.