Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GP6 Monoclonal Antibody (HY101), eBioscience™
GREENER_CHOICE

Catalog No. 14981381
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-981-381 50 μg
14-981-337 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-981-381 Supplier Invitrogen™ Supplier No. 14981381
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The HY101 monoclonal antibody reacts with the glycoprotein VI (GPVI), a 62-kDa type-I transmembrane glycoprotein that is expressed on mature megakaryotes and platelets. GPVI associates with the Fc receptor (FcR) gamma-chain to form a high affinity receptor for collagen. GPVI has also been reported to bind convulxin (Cvx). Phosphorylation of the ITAMs present on the FcR gamma-chain leads to recruitment of Src family kinases and activation of downstream signal transduction. GPVI has been reported to play a critical role in platelet activation and aggregation. Applications Reported: This HY101 antibody has been reported for use in flow cytometric analysis, and western blotting. Applications Tested: This HY101 antibody has been tested by flow cytometric analysis or immunoblotting. In flow cytometry this can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Glycoprotein VI (GP6) is a 58-kD platelet membrane glycoprotein that plays a crucial role in the collagen-induced activation and aggregation of platelets. Upon injury to the vessel wall and subsequent damage to the endothelial lining, exposure of the subendothelial matrix to blood flow results in deposition of platelets. Collagen fibers are the most thrombogenic macromolecular components of the extracellular matrix, with collagen types I, III, and VI being the major forms found in blood vessels. Platelet interaction with collagen occurs as a 2-step procedure: (1) the initial adhesion to collagen is followed by (2) an activation step leading to platelet secretion, recruitment of additional platelets, and aggregation. In physiologic conditions, the resulting platelet plug is the initial hemostatic event limiting blood loss. However, exposure of collagen after rupture of atherosclerotic plaques is a major stimulus of thrombus formation associated with myocardial infarction or stroke.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GP6
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone HY101
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene GP6
Gene Accession No. Q9HCN6
Gene Alias 9830166G18Rik; BDPLT11; Glycoprotein 5; glycoprotein 6; glycoprotein 6 (platelet); glycoprotein VI (platelet); glycoprotein VI platelet; Gm469; GP6; GPIV; GPVI; platelet collagen receptor; platelet glycoprotein VI
Gene Symbols GP6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51206
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.