Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPAA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542863 Supplier Invitrogen™ Supplier No. PA542863
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LGSLFLWREL QEAPLSLAEG WQLFLAALAQ GVLEHHTYGA LLFPLLSLGL Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%.

Posttranslational glycosylphosphatidylinositol (GPI) anchor attachment serves as a general mechanism for linking proteins to the cell surface membrane. The protein encoded by this gene presumably functions in GPI anchoring at the GPI transfer step. The mRNA transcript is ubiquitously expressed in both fetal and adult tissues. The anchor attachment protein 1 contains an N-terminal signal sequence, 1 cAMP- and cGMP-dependent protein kinase phosphorylation site, 1 leucine zipper pattern, 2 potential N-glycosylation sites, and 8 putative transmembrane domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPAA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GPAA1
Gene Accession No. O43292
Gene Alias anchor attachment protein 1; anchor attachment protein 1 (Gaa1p, yeast) homolog; C80044; GAA1; GAA1 protein homolog; glycophosphatidylinositol anchor attachment 1; glycosylphosphatidylinositol anchor attachment 1; glycosylphosphatidylinositol anchor attachment 1 (GPAA1); glycosylphosphatidylinositol anchor attachment 1 protein; glycosylphosphatidylinositol anchor attachment protein 1 homolog; Gpaa1; GPAA1 protein; GPAA1P anchor attachment protein 1 homolog; GPI anchor attachment protein 1; GPI transamidase subunit; hGAA1; mGAA1
Gene Symbols GPAA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GPAA1 (aa 558-607).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8733
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.