Learn More
Invitrogen™ GPBB Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Mouse brain. Predicted reactivity: Mouse (92%), Rat (91%), Zebrafish (85%), Chicken (86%), Sheep (92%), Rhesus Monkey (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | GPBB |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.36 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | PYGB |
| Gene Accession No. | P11216, Q8CI94 |
| Gene Alias | brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB |
| Gene Symbols | PYGB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.