Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPCR/G-protein complex-stabilizing scFv Recombinant Mouse Monoclonal Antibody (scFv16)

Catalog No. 7039761MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7039761MG 1 mg
70-397-6 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7039761MG Supplier Invitrogen™ Supplier No. 7039761MG
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Recombinant Monoclonal Antibody

The scFv antibody stabilizes GPCR/G-protein complexes for cryo-EM of active structures. It recognizes an interface between G alpha and G beta/gamma subunits, and confers resistance to nucleotide exchange-triggered dissociation of the heterotrimeric G-protein. In addition to members of the Gi/o family, this antibody fragment can also be applied to chimeric variants and stabilize other G-protein subtypes including Gs complexes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPCR/G-protein complex-stabilizing scFv
Applications Not Available
Classification Recombinant Monoclonal
Clone scFv16
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation 25mM HEPES with 10% glycerol and no preservative; pH 7.5
Host Species Mouse
Immunogen Rhodopsin/Gi1.
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.