Learn More
GPCR LS19072 Polyclonal Antibody, Invitrogen™
Description
PA1-29443 detects GPCR LS19072 from human samples.
Specifications
Specifications
| Antigen | GPCR LS19072 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | GPR135 |
| Gene Accession No. | Q8IZ08 |
| Gene Symbols | GPR135 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.