Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112160 Supplier Invitrogen™ Supplier No. PA5112160
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPD1 Polyclonal Antibody for Western Blot, IP

This gene encodes a member of the NAD-dependent glycerol-3-phosphate dehydrogenase family. The encoded protein plays a critical role in carbohydrate and lipid metabolism by catalyzing the reversible conversion of dihydroxyacetone phosphate (DHAP) and reduced nicotine adenine dinucleotide (NADH) to glycerol-3-phosphate (G3P) and NAD+. The encoded cytosolic protein and mitochondrial glycerol-3-phosphate dehydrogenase also form a glycerol phosphate shuttle that facilitates the transfer of reducing equivalents from the cytosol to mitochondria. Mutations in this gene are a cause of transient infantile hypertriglyceridemia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPD1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Gpd1
Gene Accession No. P21695
Gene Alias AI747587; Gdc1; Gdc-1; glycerol 3-phosphate dehydrogenase; glycerol phosphate dehydrogenase 1, cytoplasmic adult; Glycerol-3-phosphate dehydrogenase [NAD(+)], cytoplasmic; glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic; glycerol-3-phosphate dehydrogenase 1; glycerol-3-phosphate dehydrogenase 1 (soluble); glycerolphosphate dehydrogenase 1, cytoplasmic adult; glycerophosphate dehydrogenase; Gpd1; Gpd3; GPD-C; GPDH; GPDH-C; HTGTI; Kiaa4010; mKIAA4010
Gene Symbols Gpd1
Host Species Rabbit
Immunogen Recombinant Human GPD1/Glycerolphosphate Dehydrogenase 1 protein.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2819
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.