Learn More
Abcam GppNHp, Non-hydrolyzable GTP analog
Supplier:  Abcam AB14665910MG
Encompass_Preferred
MW 546 Da, Purity >95%. Non-hydrolyzable GTP analog. Has an amine group in place of an oxygen atom. Potent adenylate cyclase stimulator. Binds tightly to G-protein in the presence of Mg²+. Forms stable Ras complexes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1267015
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.