Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR 151 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982926
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2926 Supplier Bioss Supplier No. BS13505R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR151 is an orphan receptor.

Specifications

Antigen GPR 151
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPR151
Gene Accession No. Q8TDV0
Gene Alias AXOR40; C130082O03Rik; G protein-coupled receptor 151; Galanin receptor 4; Galanin-receptor-like protein; GALR4; GALRL; GPCR; GPCR-2037; GPR151; G-protein coupled receptor 151; G-protein coupled receptor 151 protein; G-protein coupled receptor PGR7; HGPRBMY7; nGPCR-2037; PGR7; Probable G-protein coupled receptor 151; probable G-protein coupled receptor 151 protein; putative G protein-coupled receptor nGPCR-2037; putative G-protein coupled receptor
Gene Symbols GPR151
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR 151.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134391
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.