Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551510 Supplier Invitrogen™ Supplier No. PA551510
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMEAKDGSLK AKEGSTGTSE SSVEARGSEE VRESSTVASD GSMEGKEGST KVEENSMKAD KGRTEVNQCS IDLGEDDMEF GEDDINFSED DVEAVNIPES LPPSRRNSNS NPPLPRCYQC KAA Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 57%.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. Expression of GPR101 has been documented by Northern analysis in human hypothalamus, caudate, and putamen but not in thalamus, hippocampus, frontal cortex, or pons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR101
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR101
Gene Accession No. Q96P66
Gene Alias G protein-coupled receptor 101; G protein-coupled receptor GPR101; Gm365; GPCR6; Gpr101; PAGH2; Probable G-protein coupled receptor 101; RGD1564196
Gene Symbols GPR101
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR101. Recombinant protein control fragment (Product #RP-91965).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.