Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR109B Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675238
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-238 Supplier Invitrogen™ Supplier No. PA141622
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-41622 detects GPR109B in chimpanzee and human samples. This antibody is predicted to react with canine and hamster based on sequence homology. The amino acid sequence used as immunogen is 86% in Rhesus Macaque.

Receptor for 3-OH-octanoid acid mediates a negative feedback regulation of adipocyte lipolysis to counteract prolipolytic influences under conditions of physiological or pathological increases in beta-oxidation rates. Acts as a low affinity receptor for nicotinic acid. This pharmacological effect requires nicotinic acid doses that are much higher than those provided by a normal diet. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR109B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene HCAR3
Gene Accession No. P49019
Gene Alias G protein-coupled receptor 109B; GPR109B; G-protein coupled receptor 109B; G-protein coupled receptor HM74; G-protein coupled receptor HM74B; GTP-binding protein; HCA3; HCAR3; HM74; HM74B; hydroxycarboxylic acid receptor 3; hydroxy-carboxylic acid receptor 3; Niacin receptor 2; NIACR2; Nicotinic acid receptor 2; PUMAG; Puma-g; putative chemokine receptor
Gene Symbols HCAR3
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR109B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8843
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.