Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR116 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102084 Supplier Invitrogen™ Supplier No. PA5102084
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR116.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR116 (G protein-coupled receptor 116) is a 1,346 amino acid multi-pass membrane protein that contains one SEA domain, one GPS domain and three Ig-like domains and belongs to the GPR family. Existing as a dilsulfide-liked homodimer at the cell surface, GPR116 exists as multiple alternatively spliced isoforms and is thought to play a role in regulating and maintaining proper acid-base balance throughout the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR116
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRF5
Gene Accession No. G5E8Q8, Q8IZF2
Gene Alias 8430401C09Rik; 9330185D23; ADGRF5; Adhesion G protein-coupled receptor F5; AI528491; DKFZp564O1923; FLJ90640; G protein-coupled hepta-helical receptor Ig-Hepta; G protein-coupled receptor 116; Gpr116; Gprhep; G-protein coupled hepta-helical receptor Ig-hepta; G-protein coupled receptor 116; Ig-Hepta; Ig-Hepta homolog; KIAA0758; KPG_001; mKIAA0758; probable G-protein coupled receptor 116
Gene Symbols ADGRF5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRF5(Accession Q8IZF2), corresponding to amino acid residues T1126-A1176.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221395, 224792
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.