Learn More
Invitrogen™ GPR12 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Mouse, Panda, Dog, Pig (94%).
Specifications
Specifications
| Antigen | GPR12 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | GPR12 |
| Gene Accession No. | P47775 |
| Gene Alias | FLJ18149; FLJ97704; G protein-coupled receptor 12; GPCR01; GPCR12; Gpcr20; Gpcr21; Gpr12; G-protein coupled receptor 12; MGC138349; MGC138351; PPP1R84; protein phosphatase 1, regulatory subunit 84; R334 |
| Gene Symbols | GPR12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.