Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR128 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528993
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528993 Supplier Thermo Scientific Supplier No. PA528993

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR128
Applications Western Blot
Classification Polyclonal
Concentration 3.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ADGRG7
Gene Accession No. Q96K78
Gene Alias 9130020O16Rik; Adgrg7; adhesion G protein-coupled receptor G7; adhesion G-protein coupled receptor G7; FLJ14454; FLJ16382; FLJ29035; G protein-coupled receptor 128; GPR128; G-protein coupled receptor 128; MGC142011; MGC163260; probable G-protein coupled receptor 128
Gene Symbols ADGRG7
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 1 and 355 of Human GPR128
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.