Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR128 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7210331 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4539 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4539 Supplier Proteintech Group Inc Supplier No. 247841AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.

Specifications

Antigen GPR128
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ADGRG7
Gene Accession No. Q96K78
Gene Alias G protein coupled receptor 128, GPR128
Gene Symbols ADGRG7
Host Species Rabbit
Immunogen GPR128 Fusion Protein Ag20477
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84873
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.