Learn More
Invitrogen™ GPR143 Polyclonal Antibody

Description
Immunogen sequence: CSLGFQSPRK EIQWESLTTS AAEGAHPSPL MPHENPASGK VSQVGGQTSD EALSMLSEGS DASTIEIHTA SESCNKNEGD PALPT Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.
Specifications
Specifications
| Antigen | GPR143 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GPR143 |
| Gene Accession No. | P51810 |
| Gene Alias | G protein-coupled receptor 143; GPR143; G-protein coupled receptor 143; homolog of human ocular albinism 1 (Nettleship-Falls); MOA1; NYS6; OA1; ocular albinism 1; ocular albinism type 1 protein; Ocular albinism type 1 protein homolog; RGD1565799 |
| Gene Symbols | GPR143 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.