Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR156 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555928 Supplier Invitrogen™ Supplier No. PA555928
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPDLPEQWQL WPPVPSGCAS LSSQHSYFDT ESSSSDEFFC RCHRPYCEIC FQSSSDSSDS GTSDTDPEPT GGLASWEKLW ARSKPIVNFK DDLKPTLV Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 74%.

G protein-coupled receptors (GPCRs) are a large superfamily of cell surface receptors characterized by 7 helical transmembrane domains, together with N-terminal extracellular and C-terminal intracellular domains.[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR156
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR156
Gene Accession No. Q8NFN8
Gene Alias G protein-coupled receptor 156; G protein-coupled receptor PGR28; GABABL; GABAB-related G-protein coupled receptor; GPR156; G-protein coupled receptor PGR28; PGR28; Probable G-protein coupled receptor 156
Gene Symbols GPR156
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR156. Recombinant protein control fragment (Product #RP-95163).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.