Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3005 Supplier Invitrogen™ Supplier No. PA3005
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

IHC (P) analysis shows positive staining of GPR17 in human kidney tissue.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR17 is an orphan G protein-coupled receptor involved in orchestration of oligodendrocyte differentiation and myelination in the central nervous system. GPR17 is expressed in brain, kidney, heart and umbilical vein endothelial cells. GPR17 has dual specificity for uracil nucleotides and cysteinyl leukotrienes and signals through G(i) and inhibition of adenylyl cyclase. May mediate brain damage by nucleotides and cysteinyl leukotrienes following ischemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene GPR17
Gene Accession No. Q13304
Gene Alias AI853548; DKFZp686M18273; G protein-coupled receptor 17; Gpr17; gpr17 {ECO:0000312; G-protein coupled receptor 17; P2Y-like receptor; R12; RGD:1589785}; UDP/CysLT receptor; Uracil nucleotide/cysteinyl leukotriene receptor; Y12546; Z94155
Gene Symbols GPR17
Host Species Rabbit
Immunogen KLH conjugated Cys-SFEGKTNESSLSAKSEL.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2840
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.