Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR171 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132573
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132573 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132573 Supplier Thermo Scientific Supplier No. PA132573

Rabbit Polyclonal Antibody

PA1-32573 detects Platelet activating receptor H964 from human samples.

GPR171 is a protein coding gene. Orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR171
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR171
Gene Accession No. O14626
Gene Alias A630006E05; F730001G15Rik; G protein-coupled receptor 171; Gpr171; G-protein coupled receptor H963; H963; platelet activating receptor homolog; probable G-protein coupled receptor 171
Gene Symbols GPR171
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 2nd extracellular domain of human GPR171
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.