Learn More
Invitrogen™ GPR173 Polyclonal Antibody

Description
GPR173 Polyclonal Antibody detects endogenous levels of GPR173 protein.
Specifications
Specifications
| Antigen | GPR173 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | GPR173 |
| Gene Accession No. | Q6PI62, Q9JJH2, Q9NS66 |
| Gene Alias | 3230401K02Rik; G protein-coupled receptor 173; G protein-coupled receptor SREB3; GPR173; G-protein coupled receptor 173; Probable G-protein coupled receptor 173; RP1-290F12.1; SREB3; Super conserved receptor expressed in brain 3 |
| Gene Symbols | GPR173 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.