Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3006 Supplier Invitrogen™ Supplier No. PA3006
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IHC (P) analysis shows positive staining of GPR18 in human enteric ganglion tissue. WB analysis shows GPR18 in glycoprotein-enriched fractions from GPR18 overexpressing 293 cells. A band of similar MW was also detected in GPR3 overexpressing 293 cells, suggesting some cross-reactivity with GPR3 or enhanced detection of basal GPR18 in the enriched fractions. While GPR18 has a theoretical MW of ~ 38kD, the observed band in a WB runs at ~70kD due to enrichment of the glycosylated receptor.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR18 is an orphan G protein-coupled receptor found at high levels in the testis, small intestines and cells associated with the immune system including lymphocytes, thymus and spleen. GPR18 has been found to be a receptor for endogenous lipid neurotransmitters, several of which also bind to cannabinoid receptors. Some ligands described to be GPR18 agonists or partial agonists include N-Arachidonoylglycine, Abnormal cannabidiol, O-1602, AM-251, Cannabidiol, delta 9-Tetrahydrocannabinol, N-arachidonoyl ethanolamine and Arachidonylcyclopropylamide. Antagonists include the synthetic cannabidiol related compounds CID-85469571 and O-1918.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR18
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene GPR18
Gene Accession No. Q14330
Gene Alias EMBL:AAI28783.1}; G protein-coupled receptor 18; GPCRW; Gpr18; gpr18 {ECO:0000312; G-protein coupled receptor 18; NAGly receptor; N-arachidonyl glycine receptor; RP11-461N23.8
Gene Symbols GPR18
Host Species Rabbit
Immunogen KLH conjugated Cys-RKSFRSGSLRSLSNINSEML.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2841
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.