Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532809
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532809 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532809 Supplier Invitrogen™ Supplier No. PA532809
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%); Marmoset, Bat, Elephant, Horse (94%); Bovine, Rabbit, Pig (89%); Mouse, Rat, Pufferfish (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR18 is an orphan G protein-coupled receptor found at high levels in the testis, small intestines and cells associated with the immune system including lymphocytes, thymus and spleen. GPR18 has been found to be a receptor for endogenous lipid neurotransmitters, several of which also bind to cannabinoid receptors. Some ligands described to be GPR18 agonists or partial agonists include N-Arachidonoylglycine, Abnormal cannabidiol, O-1602, AM-251, Cannabidiol, delta 9-Tetrahydrocannabinol, N-arachidonoyl ethanolamine and Arachidonylcyclopropylamide. Antagonists include the synthetic cannabidiol related compounds CID-85469571 and O-1918.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR18
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR18
Gene Accession No. Q14330
Gene Alias EMBL:AAI28783.1}; G protein-coupled receptor 18; GPCRW; Gpr18; gpr18 {ECO:0000312; G-protein coupled receptor 18; NAGly receptor; N-arachidonyl glycine receptor; RP11-461N23.8
Gene Symbols GPR18
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human GPR18.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2841
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.