Learn More
Description
A suggested positive control is EL4 cell lysate. PA5-20751 can be used with blocking peptide PEP-0865.
Specifications
Specifications
| Antigen | GPR3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GPR3 |
| Gene Accession No. | P35413, P46089, Q8K1Q3 |
| Gene Alias | ACCA; ACCA orphan receptor; adenylate cyclase constitutive activator; G protein-coupled receptor 3; G protein-coupled receptor R4; Gpcr20; Gpcr21; Gpcr3; GPR3; G-protein coupled receptor 3; G-protein-coupled receptor R4; Hacca; OTTHUMP00000043209 |
| Gene Symbols | GPR3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
