Learn More
Cayman Chemical G-15, 1161002-05-6, 1 mg
SDP
Supplier:  Cayman Chemical 146731
Encompass_Preferred
Molecular Formula C19H16BrNO2, Purity ≥95%, Formula Weight 370.2; A cell-permeable non-steroidal antagonist of the estrogen receptor GPER (Ki = 20 nM); displays low affinity cross-reactivity with the classical estrogen receptor, ER, so that at doses greater than 1 M it is capable of mediating limited ER-dependent transcriptional activity.
Catalog No. NC1215471
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.