Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100956 Supplier Invitrogen™ Supplier No. PA5100956
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR33.

Orphan receptor; could be a chemoattractant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR33
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR33
Gene Accession No. Q49SQ1
Gene Alias G protein-coupled receptor 33 (gene/pseudogene); G protein-coupled receptor 33 pseudogene; GPR33; Probable G-protein coupled receptor 33
Gene Symbols GPR33
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR33(Accession Q49SQ1), corresponding to amino acid residues I148-R198.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2856, 299007
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.