Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR34 Monoclonal Antibody (419859)
GREENER_CHOICE

Catalog No. PIMA524155
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524155 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524155 Supplier Invitrogen™ Supplier No. MA524155
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody stains human GPR34 transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.

G protein-coupled receptors (GPCRs), such as GPR34, are integral membrane proteins containing 7 putative transmembrane domains (TMs). These proteins mediate signals to the interior of the cell via activation of heterotrimeric G proteins that in turn activate various effector proteins, ultimately resulting in a physiologic response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR34
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 419859
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Gpr34
Gene Accession No. Q9UPC5
Gene Alias G protein-coupled receptor 34; Gpr34; G-protein-coupled receptor; G-protein-coupled receptor GPR34; Lypsr1; probable G-protein coupled receptor 34
Gene Symbols Gpr34
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR34.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2857
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.